Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001174054.1 | NP_001167525.1 | 16, 17 | 1340 | |
NM_007595.5 | NP_031621.3 | 16, 17 | 1379 | ||
GenBank mRNA | AK046454.1 | - | 16, 17 | 1380 | |
AK163698.1 | - | 16, 17 | 1196 | ||
BC024803.1 | - | 16, 17 | 1156 | ||
X63615.1 | - | 16, 17 | 1194 |