Gene Transcripts

Gene Symbol
Nme7
Entrez Gene ID
171567
Gene Name
NME/NM23 family member 7
Gene Aliases
D530024H21Rik, Nm23-M7, Nm23-r7
Chromosome Location
Chr. 1: 164307662 - 164437300 on Build GRCm38
UniGene ID
Mm.219428
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293313.1 NP_001280242.1 9 1074
NM_138314.4 NP_612187.2 9 1077
NM_178071.6 NP_835172.1 9 1074
GenBank mRNA AF202048.1 - 8 978
AK044962.1 - 9 1048
AK085222.1 - 9 1022
AK133221.1 - 1545
AK144915.1 - 9 1078
AK164652.1 - 9 1022
BC038021.1 - 10 1104 3963844
BC049225.1 - 8 1005 3483725
BU609451.1 - 6 661
CD775820.1 - 6 672

View Full Product Details