Gene Transcripts

Gene Symbol
Pja1
Entrez Gene ID
18744
Gene Name
praja ring finger 1, E3 ubiquitin protein ligase
Gene Aliases
praja-1
Chromosome Location
Chr. X: 99465734 - 99471268 on Build GRCm38
UniGene ID
Mm.8211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083110.2 NP_001076579.1 2 1369
NM_001290555.1 NP_001277484.1 2 1655
NM_001290556.1 NP_001277485.1 3 829
NM_008853.3 NP_032879.2 3 820
GenBank mRNA AF335250.2 - 2 1338
AK005373.1 - 2 1276
AK135382.1 - 2 1354
AK145943.1 - 2 1612
BC008148.1 - 1 643
BC025975.1 - 2 1317 3983750
BC037616.1 - 2 1292 5401000

View Full Product Details