Gene Transcripts

Gene Symbol
Rhox11
Entrez Gene ID
194738
Gene Name
reproductive homeobox 11
Gene Aliases
BC049711, Gm39
Chromosome Location
Chr. X: 38076598 - 38085139 on Build GRCm38
UniGene ID
Mm.233621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198598.2 NP_941000.1 1 411
GenBank mRNA BC049711.1 - 1 413
DQ058648.1 - 1 411

View Full Product Details