Gene Transcripts

Gene Symbol
Cc2d1a
Entrez Gene ID
212139
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
BC016188, Freud-1, Tape
Chromosome Location
Chr. 8: 84132828 - 84147931 on Build GRCm38
UniGene ID
Mm.41424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145970.1 NP_666082.2 12, 13 1375
GenBank mRNA BC016188.1 - 12, 13 1375
BC027028.1 - 11, 12 1190 5363547

View Full Product Details