Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114660.2 | NP_001108132.1 | 2 | 980 | |
NM_001286437.1 | NP_001273366.1 | 2 | 980 | ||
NM_178672.7 | NP_848787.2 | 2 | 980 | ||
GenBank mRNA | AK034000.1 | - | 2 | 975 | |
AK036637.1 | - | 2 | 922 | ||
AK044408.1 | - | 2 | 959 | ||
AK047026.1 | - | 2 | 616 | ||
AK047738.1 | - | 2 | 923 | ||
AK050673.1 | - | 2 | 931 | ||
AK054548.1 | - | 2 | 939 | ||
AK088353.1 | - | 2 | 866 | ||
AK089011.1 | - | 2 | 681 | ||
AK135666.1 | - | 2 | 216 | ||
AK140246.1 | - | 2 | 981 | ||
AK143615.1 | - | 2 | 960 | ||
AK153621.1 | - | 2 | 981 | ||
AK163028.1 | - | 2 | 939 | ||
BC042528.1 | - | 2 | 991 | 3256631 |