Gene Transcripts

Gene Symbol
Akap11
Entrez Gene ID
219181
Gene Name
A kinase (PRKA) anchor protein 11
Gene Aliases
6330501D17Rik, AKAP220, Gm80, PRKA11, mKIAA0629
Chromosome Location
Chr. 14: 78492246 - 78536982 on Build GRCm38
UniGene ID
Mm.89413
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164503.1 NP_001157975.1 7 893
GenBank mRNA AK158300.1 - 7 814

View Full Product Details