Gene Transcripts
- Gene Symbol
- Zfy2
- Entrez Gene ID
- 22768
- Gene Name
- zinc finger protein 2, Y linked
- Gene Aliases
- Zfy-2
- Chromosome Location
- Chr. Y: 2106015 - 2170409 on Build GRCm38
- UniGene ID
- Mm.411381
- Species
- Mus musculus
- Gene Symbol
- Zfy1
- Entrez Gene ID
- 22767
- Gene Name
- zinc finger protein 1, Y linked
- Gene Aliases
- Zfy-1
- Chromosome Location
- Chr. Y: 725207 - 797409 on Build GRCm38
- UniGene ID
- Mm.4324
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X14382.1 |
- |
5 |
832 |
|
View Full Product Details