Gene Transcripts

Gene Symbol
Atpaf1
Entrez Gene ID
230649
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Gene Aliases
6330547J17Rik, AI593252, ATP11, ATP11p
Chromosome Location
Chr. 4: 115784814 - 115812314 on Build GRCm38
UniGene ID
Mm.109600
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181040.4 NP_851383.2 5, 6 753
GenBank mRNA AK032020.1 - 355
BC044874.1 - 5, 6 594 4487325
CO431467.1 - 5, 6 340

View Full Product Details