Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172146.1 | NP_001165617.1 | 2 | 384 | |
NM_146165.2 | NP_666277.1 | 2 | 264 | ||
GenBank mRNA | BC024410.1 | - | 2 | 357 | |
BC024480.1 | - | 2 | 255 | 4977193 | |
BC026958.1 | - | 2 | 323 | 4013019 | |
BC026972.1 | - | 2 | 337 | 5371230 | |
CF732613.1 | - | 2 | 306 | ||
CJ051555.1 | - | 2 | 256 |