Gene Transcripts

Gene Symbol
Adamts17
Entrez Gene ID
233332
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 17
Gene Aliases
AU023434
Chromosome Location
Chr. 7: 66839723 - 67153171 on Build GRCm38
UniGene ID
Mm.445763
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033877.4 NP_001029049.2 9, 10 1410
GenBank mRNA AK053131.1 - 9, 10 1411
BC141170.1 - 9, 10 1362
CF741491.1 - 1, 2 152
CX769149.1 - 1, 2 113

View Full Product Details