Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_183020.1 | NP_898841.1 | 6 | 742 | |
GenBank mRNA | AK084800.1 | - | 6 | 814 | |
AK155062.1 | - | 6 | 807 | ||
AK158626.1 | - | 6 | 429 | ||
AK168745.1 | - | 6 | 459 | ||
BC054483.1 | - | 6 | 742 | 6391591 |