Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177700.4 | NP_808368.3 | 4 | 1790 | |
GenBank mRNA | AK087334.1 | - | 1 | 1101 | |
AK122281.1 | - | 4 | 1672 | ||
AX775769.1 | - | 4 | 1790 | ||
BC025922.1 | - | 1 | 396 | ||
BC058974.1 | - | 3 | 1335 | ||
BC060631.1 | - | 4 | 1775 | 6847850 |