Gene Transcripts

Gene Symbol
Aadat
Entrez Gene ID
23923
Gene Name
aminoadipate aminotransferase
Gene Aliases
AI875679, Aadt, KATII, Kat2, mKat-2
Chromosome Location
Chr. 8: 60505870 - 60545677 on Build GRCm38
UniGene ID
Mm.35020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011834.2 NP_035964.1 7, 8 896
GenBank mRNA AF072376.1 - 7, 8 896
AK075578.1 - 7, 8 902
AK144042.1 - 7, 8 849
BC012637.1 - 7, 8 899 4160508

View Full Product Details