Gene Transcripts

Gene Symbol
Sh3yl1
Entrez Gene ID
24057
Gene Name
Sh3 domain YSC-like 1
Gene Aliases
AI314953, Ray, YSC84
Chromosome Location
Chr. 12: 30911669 - 30960160 on Build GRCm38
UniGene ID
Mm.218624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289480.1 NP_001276409.1 6 1039
NM_013709.5 NP_038737.1 7 1153
GenBank mRNA AK141199.1 - 7 1154
AK168483.1 - 8 779
BC010296.1 - 6 601 3154966
BC058557.1 - 6 1016 6313231
BC110043.1 - 7 1131 5149633
D85926.1 - 7 636

View Full Product Details