Gene Transcripts

Gene Symbol
Tll2
Entrez Gene ID
24087
Gene Name
tolloid-like 2
Gene Aliases
Chromosome Location
Chr. 19: 41080940 - 41210765 on Build GRCm38
UniGene ID
Mm.89983
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011904.3 NP_036034.1 16 2189
GenBank mRNA AF073526.1 - 16 2250
BC132537.1 - 16 2189 40130814
BC145132.1 - 16 2138 9053680

View Full Product Details