Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011907.4 | NP_036037.1 | 2 | 311 | |
GenBank mRNA | AF151108.1 | - | 2 | 298 | |
BC104384.1 | - | 1 | 276 | 40050180 | |
BC104385.2 | - | 1 | 276 | 40050181 | |
BY714289.1 | - | 2 | 313 |