Gene Transcripts

Gene Symbol
Xrn2
Entrez Gene ID
24128
Gene Name
5'-3' exoribonuclease 2
Gene Aliases
Chromosome Location
Chr. 2: 147012631 - 147078002 on Build GRCm38
UniGene ID
Mm.3065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011917.2 NP_036047.2 27 2553
GenBank mRNA AK004800.1 - 27 2528
AK011711.1 - 27 2557
AK031247.1 - 27 2596
AK053643.1 - 27 2546
AK167019.1 - 27 2573
AK168037.1 - 27 2599
BC004028.1 - 19 1724 3495063
BC052395.1 - 16 1469
BC052823.1 - 27 2414
BC054743.1 - 27 2553 6400974
D38517.1 - 27 2491

View Full Product Details