Gene Transcripts

Gene Symbol
Bsx
Entrez Gene ID
244813
Gene Name
brain specific homeobox
Gene Aliases
Bsx1a, Bsx1b
Chromosome Location
Chr. 9: 40871854 - 40878962 on Build GRCm38
UniGene ID
Mm.261652
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178245.3 NP_839976.1 1 260
GenBank mRNA AY233396.1 - 1 260
BC104386.1 - 1 258 40050191
BC104387.1 - 1 258 40050192

View Full Product Details