Gene Transcripts

Gene Symbol
Adh4
Entrez Gene ID
26876
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Gene Aliases
Adh2
Chromosome Location
Chr. 3: 138415497 - 138430892 on Build GRCm38
UniGene ID
Mm.158750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011996.2 NP_036126.2 2, 3 145
GenBank mRNA AJ245750.1 - 2, 3 133
AK132994.1 - 2, 3 145
BC100729.1 - 2, 3 133 40045447
BC100730.1 - 2, 3 132 40045448
BC100736.1 - 2, 3 133 40045445

View Full Product Details