Gene Transcripts

Gene Symbol
Vat1
Entrez Gene ID
26949
Gene Name
vesicle amine transport protein 1 homolog (T californica)
Gene Aliases
VAT-1
Chromosome Location
Chr. 11: 101458748 - 101466199 on Build GRCm38
UniGene ID
Mm.29210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012037.2 NP_036167.1 5, 6 1207
GenBank mRNA AK034795.1 - 5, 6 1209
AK145193.1 - 5, 6 1241
AK154965.1 - 5, 6 1190
AK155342.1 - 5, 6 1209
AK159310.1 - 5, 6 1150
AK159315.1 - 5, 6 1192
AK160891.1 - 5, 6 1199
AK170371.1 - 5, 6 1188
AK216339.1 - 1, 2 196
BC024586.1 - 3, 4 436
BC025496.1 - 5, 6 1163 5320239
BC036169.1 - 4, 5 649
BC085504.1 - 5, 6 1126
BC099678.1 - 5, 6 1189 6311681

View Full Product Details