Gene Transcripts

Gene Symbol
Anks1b
Entrez Gene ID
77531
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA-1b, C030032C09Rik, E530015N03Rik, ENSMUSG00000078431, Gm10937, Gm1555
Chromosome Location
Chr. 10: 89873509 - 90973300 on Build GRCm38
UniGene ID
Mm.440605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128086.2 NP_001121558.2 13 2340

View Full Product Details