Gene Transcripts

Gene Symbol
Wfikkn2
Entrez Gene ID
278507
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2
Gene Aliases
2610304F08Rik, AY100450, Gasp1, WFIKKNRP
Chromosome Location
Chr. 11: 94235951 - 94247793 on Build GRCm38
UniGene ID
Mm.342216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181819.2 NP_861540.2 2 342
GenBank mRNA AY308804.1 - 2 312
BC113144.1 - 2 312 40047070
JQ080910.1 - 2 312

View Full Product Details