Gene Transcripts

Gene Symbol
Wdr59
Entrez Gene ID
319481
Gene Name
WD repeat domain 59
Gene Aliases
5430401O09Rik, Cdw12
Chromosome Location
Chr. 8: 111448781 - 111522107 on Build GRCm38
UniGene ID
Mm.247294
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170742.1 NP_001164213.1 18 1860
NM_001170743.1 NP_001164214.1 18 1860
NM_176923.4 NP_795897.2 18 1860
GenBank mRNA AK030644.1 - 18 1741
AK040807.1 - 3 147
AK136303.1 - 14 1412
AK170910.1 - 18 1859
AK173301.1 - 18 1823
BC066082.1 - 18 1845 30355918
EF011622.1 - 18 1845

View Full Product Details