Gene Transcripts

Gene Symbol
Smo
Entrez Gene ID
319757
Gene Name
smoothened homolog (Drosophila)
Gene Aliases
E130215L21Rik, Smoh, bnb, smoothened
Chromosome Location
Chr. 6: 29735497 - 29761366 on Build GRCm38
UniGene ID
Mm.29279
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176996.4 NP_795970.3 8, 9 1980
GenBank mRNA AK081050.1 - 2, 3 168
AK194911.1 - 4 372
BC096028.1 - 8, 9 1499 6850065
BQ947565.1 - 3, 4 242
CA749812.1 - 5, 6 580
FJ883552.1 - 8, 9 1489

View Full Product Details