Gene Transcripts

Gene Symbol
Lin28b
Entrez Gene ID
380669
Gene Name
lin-28 homolog B (C. elegans)
Gene Aliases
2810403D23Rik, D030047M17Rik, Lin-28.2
Chromosome Location
Chr. 10: 45376619 - 45487303 on Build GRCm38
UniGene ID
Mm.440328
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031772.2 NP_001026942.1 3, 4 522
GenBank mRNA AK133928.1 - 3, 4 541
BC089037.1 - 3, 4 513 30690977
DQ127225.1 - 3, 4 522

View Full Product Details