Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001293683.1 | NP_001280612.1 | 4 | 403 | |
NM_001293684.1 | NP_001280613.1 | 4 | 403 | ||
NM_025635.4 | NP_079911.1 | 4 | 403 | ||
GenBank mRNA | AK004305.1 | - | 4 | 355 | |
AK008144.1 | - | 4 | 353 | ||
AK013556.1 | - | 4 | 338 | ||
AK075977.1 | - | 4 | 349 | ||
AK082215.1 | - | 4 | 352 | ||
AK134380.1 | - | 4 | 355 | ||
BC013559.1 | - | 4 | 321 | ||
BC027100.1 | - | 4 | 360 | 5359154 | |
BC034870.1 | - | 4 | 332 | 1382192 |