Gene Transcripts

Gene Symbol
Zfp410
Entrez Gene ID
52708
Gene Name
zinc finger protein 410
Gene Aliases
Apa1, D12Ertd748e, Znf410
Chromosome Location
Chr. 12: 84315928 - 84344439 on Build GRCm38
UniGene ID
Mm.255150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144833.3 NP_659082.1 12 1884
GenBank mRNA AF295806.1 - 12 1646
AK051939.1 - 10 1284
AK083003.1 - 12 1627
AK132345.1 - 12 1623
BC021528.1 - 12 1639 5353673
BC056957.1 - 12 1581

View Full Product Details