Gene Transcripts

Gene Symbol
Slc2a5
Entrez Gene ID
56485
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 5
Gene Aliases
AI526984, Glut5, Slc5a
Chromosome Location
Chr. 4: 150119344 - 150144168 on Build GRCm38
UniGene ID
Mm.260220
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019741.3 NP_062715.2 4 733
GenBank mRNA AF161071.1 - 2 94
AK029720.1 - 4 734
BC023500.1 - 2 109 5043284

View Full Product Details