Gene Transcripts

Gene Symbol
Treh
Entrez Gene ID
58866
Gene Name
trehalase (brush-border membrane glycoprotein)
Gene Aliases
2210412M19Rik
Chromosome Location
Chr. 9: 44673233 - 44686305 on Build GRCm38
UniGene ID
Mm.45380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021481.3 NP_067456.1 5 528
GenBank mRNA AF136944.1 - 5 523
AK008912.1 - 5 513

View Full Product Details