Gene Transcripts

Gene Symbol
Ribc1
Entrez Gene ID
66611
Gene Name
RIB43A domain with coiled-coils 1
Gene Aliases
2610028I09Rik, W08639
Chromosome Location
Chr. X: 152004580 - 152018219 on Build GRCm38
UniGene ID
Mm.158120
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025660.2 NP_079936.1 3 282
GenBank mRNA AK011592.1 - 3 282
AK017461.1 - 3 174
BC050755.1 - 3 270 6772058

View Full Product Details