Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027654.2 | NP_081930.1 | 6, 7 | 826 | |
GenBank mRNA | AB047007.1 | - | 6, 7 | 876 | |
AK016709.1 | - | 6, 7 | 849 | ||
AK132448.1 | - | 6, 7 | 872 | ||
BC016195.1 | - | 6, 7 | 826 | 4488614 | |
BC089460.1 | - | 6, 7 | 865 | 30632537 |