Gene Transcripts

Gene Symbol
Spats2
Entrez Gene ID
72572
Gene Name
spermatogenesis associated, serine-rich 2
Gene Aliases
2700012F11Rik, 59kDa, Scr59, p59
Chromosome Location
Chr. 15: 99125926 - 99213215 on Build GRCm38
UniGene ID
Mm.276650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139140.1 NP_631879.1 8 1135
GenBank mRNA AB079132.1 - 8 1135
AK048720.1 - 8 1177
AK159335.1 - 8 1230
AK167788.1 - 7 813
AK171266.1 - 7 891
AK215485.1 - 2 149
BC023432.1 - 6 785 5029821
BC072579.1 - 8 1402

View Full Product Details