Gene Transcripts

Gene Symbol
Acsl3
Entrez Gene ID
74205
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
2610510B12Rik, Acs3, C85929, Facl3, Pro2194
Chromosome Location
Chr. 1: 78657825 - 78707743 on Build GRCm38
UniGene ID
Mm.276016
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033606.2 NP_001028778.2 2 348
NM_028817.3 NP_083093.2 3 488
GenBank mRNA AK012088.1 - 2 291
AK161708.1 - 3 489
AK166723.1 - 3 427
BC031529.1 - 1 73 4009004

View Full Product Details