Gene Transcripts

Gene Symbol
Mrgprh
Entrez Gene ID
80978
Gene Name
MAS-related GPR, member H
Gene Aliases
Gpr90, Mgrb, MrgH
Chromosome Location
Chr. 17: 12876034 - 12877842 on Build GRCm38
UniGene ID
Mm.160368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030726.1 NP_109651.1 1 1740
GenBank mRNA AF295365.1 - 1 1740
BC116696.1 - 1 1370 40105568
BC117525.1 - 1 1368 40105567

View Full Product Details