Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002238.2 | NP_001002238.1 | 7 | 816 | |
NM_001002240.2 | NP_001002240.1 | 7 | 789 | ||
NM_001002241.2 | NP_001002241.1 | 6 | 1089 | ||
NM_031387.3 | NP_113564.2 | 7 | 949 | ||
GenBank mRNA | AB067571.1 | - | 6 | 770 | |
AB183526.1 | - | 7 | 816 | ||
AB183527.1 | - | 7 | 789 | ||
AB183528.1 | - | 6 | 1089 | ||
AB183529.1 | - | 6 | 653 | ||
AF285591.1 | - | 3 | 143 | ||
AK029806.1 | - | 7 | 950 | ||
BC129954.1 | - | 2 | 75 | 40081321 | |
BC129955.1 | - | 2 | 75 | 40081323 |