Gene Transcripts

Gene Symbol
Srrt
Entrez Gene ID
83701
Gene Name
serrate RNA effector molecule homolog (Arabidopsis)
Gene Aliases
2810019G02Rik, ASR2A, ASR2B, ASR2C, ASR2D, Ars2, Asr2
Chromosome Location
Chr. 5: 137295704 - 137307714 on Build GRCm38
UniGene ID
Mm.387734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109909.1 NP_001103379.1 10, 11 1577
NM_001109910.1 NP_001103380.1 10, 11 1577
NM_031405.2 NP_113582.1 10, 11 1577
GenBank mRNA AK144300.1 - 10, 11 1569
BC019117.1 - 1, 2 119
BC066831.1 - 10, 11 1536 30471339

View Full Product Details