Gene Transcripts

Gene Symbol
Abtb2
Entrez Gene ID
99382
Gene Name
ankyrin repeat and BTB (POZ) domain containing 2
Gene Aliases
AW539457, BPOZ-2
Chromosome Location
Chr. 2: 103566310 - 103718423 on Build GRCm38
UniGene ID
Mm.35850
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178890.3 NP_849221.2 2 1324
GenBank mRNA AK042322.1 - 2 1322
AK151620.1 - 2 1320
AK152163.1 - 2 1320
AK170569.1 - 2 1323
AK172414.1 - 2 1321
AK172468.1 - 2 1321
AK189625.1 - 2 119
BC054399.1 - 2 1324 6493840

View Full Product Details