Gene Transcripts

Gene Symbol
Slc22a13
Entrez Gene ID
102570
Gene Name
solute carrier family 22 (organic cation transporter), member 13
Gene Aliases
AI648912, OCTL1, OCTL3, ORCTL3
Chromosome Location
Chr. 9: 119192968 - 119209105 on Build GRCm38
UniGene ID
Mm.38775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133980.3 NP_598741.2 1 82
GenBank mRNA AF529220.1 - 1 75
BC021449.1 - 1 74 5044564

View Full Product Details