Gene Transcripts

Gene Symbol
Mpeg1
Entrez Gene ID
17476
Gene Name
macrophage expressed gene 1
Gene Aliases
MPS1, Mpg-1
Chromosome Location
Chr. 19: 12460709 - 12465285 on Build GRCm38
UniGene ID
Mm.3999
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010821.1 NP_034951.1 2 1748
GenBank mRNA AK030216.1 - 2 1751
BC049256.1 - 1 55
BC129843.1 - 1 1642 40095576
BC129844.1 - 1 1642 40095579
L20315.1 - 1 1651

View Full Product Details