Gene Transcripts
- Gene Symbol
- Spag6l
- Entrez Gene ID
- 381350
- Gene Name
- sperm associated antigen 6 like
- Gene Aliases
- Chromosome Location
- Chr. 2: 18694011 - 18749746 on Build GRCm38
- UniGene ID
- Mm.330024
- Species
- Mus musculus
- Gene Symbol
- Gm3363
- Entrez Gene ID
- 100041479
- Gene Name
- predicted gene 3363
- Gene Aliases
- Chromosome Location
- Chr. 2: 18742709 - 18760862 on Build GRCm38
- UniGene ID
- Mm.393659
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI552988.1 |
- |
1 |
64 |
|
AK140973.1 |
- |
5 |
1260 |
|
View Full Product Details