Gene Transcripts

Gene Symbol
March11
Entrez Gene ID
211147
Gene Name
membrane-associated ring finger (C3HC4) 11
Gene Aliases
9630025C22
Chromosome Location
Chr. 15: 26308904 - 26409594 on Build GRCm38
UniGene ID
Mm.54257
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177597.5 NP_808265.2 4 1129
GenBank mRNA AK162044.1 - 3 931

View Full Product Details