Gene Transcripts

Gene Symbol
Pabpc6
Entrez Gene ID
67543
Gene Name
poly(A) binding protein, cytoplasmic 6
Gene Aliases
4932702K14Rik, AI428050, Pabpc3
Chromosome Location
Chr. 17: 9666497 - 9669704 on Build GRCm38
UniGene ID
Mm.26987
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163836.1 NP_001157308.1 1 589
GenBank mRNA AK016578.1 - 1 582
AK029765.1 - 1 570
BC113177.1 - 1 559

View Full Product Details