Gene Transcripts
- Gene Symbol
- Phykpl
- Entrez Gene ID
- 72947
- Gene Name
- 5-phosphohydroxy-L-lysine phospholyase
- Gene Aliases
- 2900006B13Rik, Agxt2l2
- Chromosome Location
- Chr. 11: 51583983 - 51603269 on Build GRCm38
- UniGene ID
- Mm.256875
- Species
- Mus musculus
- Gene Symbol
- Hnrnpab
- Entrez Gene ID
- 15384
- Gene Name
- heterogeneous nuclear ribonucleoprotein A/B
- Gene Aliases
- 3010025C11Rik, CBF-A, Cgbfa, Hnrpab
- Chromosome Location
- Chr. 11: 51600100 - 51606881 on Build GRCm38
- UniGene ID
- Mm.256875
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK047205.1 |
- |
|
697 |
|
View Full Product Details