Gene Transcripts

Gene Symbol
Celf6
Entrez Gene ID
76183
Gene Name
CUGBP, Elav-like family member 6
Gene Aliases
6330569O16Rik, Brunol6
Chromosome Location
Chr. 9: 59578286 - 59607293 on Build GRCm38
UniGene ID
Mm.265415
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175235.3 NP_780444.2 8 991
GenBank mRNA AK034216.1 - 7 787
BC052406.1 - 8 991 5716119
BC057083.1 - 7 773 5707546

View Full Product Details