Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173347.2 | NP_775482.1 | 5 | 1108 | |
GenBank mRNA | AF051908.2 | - | 5 | 655 | |
AK081637.1 | - | 5 | 799 | ||
AK142160.1 | - | 5 | 707 | ||
AK202889.1 | - | 2 | 239 | ||
BC044877.1 | - | 3 | 533 | ||
BC057546.1 | - | 5 | 1108 | 6416282 | |
BC058635.1 | - | 5 | 837 | 5698271 |