Gene Transcripts

Gene Symbol
Sec14l4
Entrez Gene ID
103655
Gene Name
SEC14-like 4 (S. cerevisiae)
Gene Aliases
AI256582, AW536553, SPF, TAP
Chromosome Location
Chr. 11: 4031378 - 4052571 on Build GRCm38
UniGene ID
Mm.337476
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146013.1 NP_666125.1 6 501
GenBank mRNA AK028954.1 - 6 462
BC026948.1 - 6 501 5136664

View Full Product Details