Gene Transcripts

Gene Symbol
Fam208b
Entrez Gene ID
105203
Gene Name
family with sequence similarity 208, member B
Gene Aliases
AI645998, Pap20, mKIAA2006
Chromosome Location
Chr. 13: 3566035 - 3611231 on Build GRCm38
UniGene ID
Mm.221609
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134063.3 NP_598824.3 14 3782
GenBank mRNA AK131707.1 - 3 1178
AK220437.1 - 14 3770
BC085160.1 - 5 2525

View Full Product Details