Gene Transcripts

Gene Symbol
Aipl1
Entrez Gene ID
114230
Gene Name
aryl hydrocarbon receptor-interacting protein-like 1
Gene Aliases
A930007I01Rik
Chromosome Location
Chr. 11: 72028717 - 72042375 on Build GRCm38
UniGene ID
Mm.95707
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053245.2 NP_444475.2 2 313
GenBank mRNA AF296412.1 - 2 262
BC028285.1 - 2 313 4501401
CK618559.1 - 2 313

View Full Product Details