Gene Transcripts

Gene Symbol
Alox12b
Entrez Gene ID
11686
Gene Name
arachidonate 12-lipoxygenase, 12R type
Gene Aliases
Aloxe2, e-LOX2
Chromosome Location
Chr. 11: 69157072 - 69169791 on Build GRCm38
UniGene ID
Mm.340329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009659.2 NP_033789.1 3 588
GenBank mRNA AF059251.1 - 3 466
BC113149.1 - 3 552 40054352
Y14334.1 - 3 588

View Full Product Details